missing translation for 'onlineSavingsMsg'
Learn More

Diethylphosphonoacetic Acid, TRC

Product Code. 30375262
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375262 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375262 Supplier Toronto Research Chemicals Supplier No. TRCD44474525G
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethylphosphonoacetic Acid
CAS 3095-95-2
Molecular Formula C6H13O5P
Quantity 25 g
Synonym (Diethoxyphosphinyl)acetic Acid, (Diethylphosphono)acetic Acid, 2-(Diethoxyphosphoryl)acetic Acid, Diethoxyphosphorylacetic Acid, Diethyl Carboxymethanephosphonate, Diethyl Carboxymethylphosphonate, NSC 272281
InChI Formula InChI=1S/C6H13O5P/c1-3-10-12(9,11-4-2)5-6(7)8/h3-5H2,1-2H3,(H,7,8)
SMILES CCOP(=O)(CC(=O)O)OCC
IUPAC Name 2-diethoxyphosphorylacetic acid
Molecular Weight (g/mol) 196.14
Formula Weight 196.0501
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.