missing translation for 'onlineSavingsMsg'
Learn More

Diethylvinylphosphonate, 97%, Thermo Scientific Chemicals

Product Code. 11305828
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11305828 10 g 10g
11315828 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11305828 Supplier Thermo Scientific Alfa Aesar Supplier No. 010368.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Diethylvinylphosphonate is used in the preparation of diethyl N-alkyl-2-aminoethylphosphonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 682-30-4
Molecular Formula C6H13O3P
Molecular Weight (g/mol) 164.14
MDL Number MFCD00009079
InChI Key DREPONDJUKIQLX-UHFFFAOYSA-N
Synonym diethyl vinylphosphonate, vinylphosphonic acid diethyl ester, diethyl vinylphosphate, diethoxyphosphinylethene, phosphonic acid, vinyl-, diethyl ester, diethyl ethenylphosphonate, phosphonic acid, ethenyl-, diethyl ester, 1-ethenyl ethoxy phosphoryl oxyethane, acmc-1at5g
PubChem CID 69629
SMILES CCOP(=O)(OCC)C=C

Specifications

Density 1.068 g/mL
Boiling Point 202°C
Flash Point 110°C (230°F)
Refractive Index 1.429
Quantity 10 g
UN Number UN3278
Beilstein EM>5
IUPAC Name 1-[ethenyl(ethoxy)phosphoryl]oxyethane
Formula Weight 164.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethylvinylphosphonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.