Learn More
BD Difco™ Bordet Gengou Agar Base
Product Code. 11748942
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. 11748942
Brand: BD 248200
Enriched with defibrinated blood for the isolation of B. pertussis and other Bordetella species. BD Difco™ Bordet Gengou Agar Base is part of a large selection of dehydrated media.
  • Enriched with defibrinated blood
  • Dehydrated
  • Culture media
TRUSTED_SUSTAINABILITY
Specifications
Media Type Dehydrated
Product Type Agar
For Use With (Application) For the detection and isolation of B. pertussis and other Bordetella species.
Quantity 500 g