missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ Casein Digest

Product Code. 30485066
Change view
Click to view available options
Quantity:
2 kg
Unit Size:
2kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30485066 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30485066 Supplier Gibco™ Supplier No. 214896

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Difco Casein Digest is an animal origin (AO) enzymatic digest of casein with different enzymatic digestion conditions than other casein.

Difco Casein Digest is an animal origin (AO) enzymatic digest of casein with different enzymatic digestion conditions than other casein digests in our portfolio, such as Bacto Casitone and Bacto Tryptone. Difco Casein Digest has a unique nutritional profile of amino acids, nutrients, minerals, nucleotide precursors, peptides, and other metabolites ideal for growth promotion in fermentation applications, especially Escherichia coli.

Features:

  • Unique nutritional profile of amino acids, nutrients, minerals, nucleotide precursors, peptides, and other metabolites

Applications of Difco Casein Digest include:

  • Microbial fermentation
  • Growth promotion, specifically E. Coli

Applications:

Cultivation of heterotrophic microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance LUO, MUO
Color Light Beige
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Additive Casein Digest
Form Free-flowing Powder
Format Pail
Grade Technical
Manufacturing Origin United States
Media Type Cell Culture Supplement
Product Type Peptone
Sterility Non-sterile
For Use With (Application) Cultivation of heterotrophic microorganisms
Product Line Difco
Quantity 2 kg
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.