missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ M-Green Yeast and Mold Broth
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
M-Green Yeast and Mold Broth is a modification of the liquid medium, M-Yeast and Mold Broth, which was developed to improve the efficiency of detection and enumeration of fungi in sugar and other materials by the membrane filter method. The revised formula contains the indicator dye, bromcresol green.
Specifications
Specifications
| Broth Type | M-Green Yeast and Mold Broth |
| Detects | Fungi |
| Form | Solid |
| For Use With (Application) | Detection of fungi in the routine analysis of beverage |
| Incubation Conditions | 30° to 35°C for 2 days |
| Purification Method | Membrane filter method |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction