missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Poly Antisera

Product Code. 11702874
Change view
Click to view available options
Product Type:
Shigella Poly Group A
Shigella Poly Group A1
Shigella Poly Group B
Shigella Poly Group D
Unit Size:
3mL
5mL
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
11702874 Shigella Poly Group A1 3mL
17352361 Shigella Poly Group B Each
11894422 Shigella Poly Group A 5mL
11772894 Shigella Poly Group D 3mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11702874 Supplier BD Supplier No. 227761
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For agglutination test

Salmonella poly antiserum

  • Febrile control antisera
  • Helps in the detection of diseases such as salmonellosis, brucellosis and rickettsial diseases
  • Qualitative and semi-quantitative slide or tube agglutination tests for homologous antibodies
  • Used along with febrile antigens as positive or negative controls
Shigella antiserum poly group
  • For identification of morphologically and biochemically identified Shigella species
  • Can be used only in slide agglutination test
  • Lyophilized, polyclonal rabbit antisera

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance For in vitro diagnostic use
Product Type Shigella Poly Group A1
Quantity 3 mL
compliance-icons
Product Identifier
  • Vial Shigella Antis Poly A1 Lyophilized
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sodium Azide

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.