missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Difco™ PPLO Broth
Click to view available options
Quantity:
10 kg
Unit Size:
10kg
Description
A complete media used in the cultivation of microorganisms from the class Mollicutes. Used in animal health applications for the propagation of Mycoplasma species such as Mycoplasma mycoides, Mycoplasma gallisepticum, and others for vaccine production.
Specifications
Specifications
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
| Type | Dehydrated culture media |
| For Use With (Application) | Isolating and cultivating Mycoplasma |
| Product Line | Difco |
| Quantity | 10 kg |
| Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction