missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ Soytone

An enzymatic digest of soy.

121.00 €

Specifications

Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Yeast Supplement
For Use With (Application) Mammalian Cell Culture
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16219781
View Documents Promotion Details
Gibco™
212488
500 g
121.00 €
500g
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • An alternative carbohydrate profile to yeast supplements for mammalian cell processes, such as CHO
  • High concentrations of vitamins and carbohydrates
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifications

Specifications

Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Powder
Mammalian Cell Culture
Animal origin-free
Mammalian, Insect, Microbial
Yeast Supplement
Difco™
Videos
SDS
Documents

Documents

Product Certifications