missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Difco™ Yeast Extract, low-dusting (LD)
Description
- Yeast extract designed to limit the amount of dust generated compared to other spray-dried yeast extracts
- Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells
- Gibco™ yeast extracts are derived from primary grown baker’s yeast
- These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
- Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation
Specifications
Specifications
| Certifications/Compliance | This product does not contains HAPs; This product does not contain any Proposition 65 chemicals; WHMIS Hazard Class - Non-controlled - This product has been classified in accordance with the hazard criteria of the Controlled Products Regulations (CPR)and the SDS contains all the information required by the CPR |
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
| Cell Type | Bacteria |
| Culture Type | Mammalian, Insect, Microbial |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| For Use With (Application) | Mammalian, Microbial, Vaccine, Insect cell culture applications |
| Product Line | Difco |
| Quantity | 10 kg |
| Source | Yeast, animal origin-free |
Product Title
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?