missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Difco Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells derived from primary grown baker's yeast
- These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
- Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation
- Difco Yeast Extract Low-Dusting (LD) was designed to limit the amount of dust generated compared to other spray-dried yeast extracts
- It is ultra-filtered and specifically designed for mammalian cell culture applications
- With its low endotoxin level and high content of naturally occurring B vitamins, it can be used effectively to reduce the need for fetal bovine serum
- It has an endotoxin level of less than or equal to 500 EU/g
- Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifications
Specifications
| Certifications/Compliance | LUO |
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
| Cell Type | Bacteria |
| Culture Type | Mammalian, Insect, Microbial |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Format | Bottle |
| For Use With (Application) | Cell culture and microbial fermentation, tissue culture application |
| Product Line | Difco |
| Quantity | 500 g |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?