missing translation for 'onlineSavingsMsg'
Learn More

Difenoconazole-d6 HCl (1,3-dioxolan-4,5,5-d3-4-methyl-d3) (mixture of diastereomers), CDN

Product Code. 30312319
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30312319

Brand: CDN CDND79230.005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Difenoconazole-D6 (4-(methyl-D3)-1,3-dioxolan-2-yl-4,5,5-D3) Hydrochloride
CAS 2748435-81-4
Molecular Formula C19 D6 H11 Cl2 N3 O3 . Cl H
Quantity 0.005 g
Synonym Difenoconazole D6 (1,1,2,3,3,3-propyl D6) hydrochloride, Difenoconazole-d6 hydrochloride, Difenoconazole-d6 HCl (1,3-dioxolan-4,5,5-d3-4-methyl-d3) (mixture of diastereomers), Difenoconazole-D6 (4-(methyl-D3)-1,3-dioxolan-2-yl-4,5,5-D3) hydrochloride, 1-[[2-[2-Chloro-4-(4-chlorophenoxy)phenyl]-4-(methyl-d3)-1,3-dioxolan-2-yl-4,5,5-d3]methyl]-1 H-1,2,4-triazole hydrochloride (1:1)
InChI Formula InChI=1 S/C19H17Cl2N3O3.ClH/c1-13-9-25-19(27-13,10-24-12-22-11-23-24)17-7-6-16(8-18(17)21)26-15-4-2-14(20)3-5-15;/h2-8,11-13 H,9-10H2,1H3;1 H/i1D3,9D2,13 D;
SMILES Cl.[2 H]C([2 H])([2 H])C1([2 H])OC(Cn2cncn2)(OC1([2 H])[2 H])c3ccc(Oc4ccc(Cl)cc4)cc3Cl
IUPAC Name 1-[[2-[2-chloro-4-(4-chlorophenoxy)phenyl]-4,4,5-trideuterio-5-(trideuteriomethyl)-1,3-dioxolan-2-yl]methyl]-1,2,4-triazole;hydrochloride
Molecular Weight (g/mol) 448.7605
Formula Weight 447.079 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.