Learn More
Diffraction Grating Spectroscope
Product Code. 13112990
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity Unit Size
13112990 1 Ea. Each
1 options
Product Code. 13112990 Supplier Brand Not Defined Supplier No. TE313
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Specifications
For Use With (Application) For demonstrating the electromagnetic spectrum and the emission and absorption phenomena
Quantity 1 Ea.
Length (Metric) 110 mm
Product Type Diffraction grating spectroscope