missing translation for 'onlineSavingsMsg'
Learn More

Difluoroacetic acid, 98%

Product Code. 11475133
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475133 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11475133 Supplier Thermo Scientific Alfa Aesar Supplier No. A12787.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 381-73-7 | C2H2F2O2 | 96.033 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 381-73-7
Molecular Formula C2H2F2O2
Molecular Weight (g/mol) 96.033
MDL Number MFCD00004220
InChI Key PBWZKZYHONABLN-UHFFFAOYSA-N
Synonym difluoroacetic acid, acetic acid, difluoro, difluoressigsaeure, 1,1-difluoroacetic acid, unii-zqk1c95k3n, zqk1c95k3n, acetic acid, 2,2-difluoro, difluoroaceticacid, difluoracetic acid, difluoro-acetic acid
PubChem CID 9788
ChEBI CHEBI:23716
IUPAC Name 2,2-difluoroacetic acid
SMILES C(C(=O)O)(F)F

Specifications

Melting Point -1°C
Density 1.526
Boiling Point 132°C to 134°C
Flash Point 78°C (172°F)
Odor Pungent
Refractive Index 1.343
Quantity 25 g
UN Number UN3265
Beilstein 1098588
Formula Weight 96.03
Percent Purity 98%
Chemical Name or Material Difluoroacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.