missing translation for 'onlineSavingsMsg'
Learn More

Difluoroacetic acid, 98%

Product Code. 10532761 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 mL
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
2.5mL
50mL
This item is not returnable. View return policy

Product Code. 10532761

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 381-73-7 | C2H2F2O2 | 96.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 381-73-7
Molecular Formula C2H2F2O2
Molecular Weight (g/mol) 96.03
MDL Number MFCD00004220
InChI Key PBWZKZYHONABLN-UHFFFAOYSA-N
Synonym difluoroacetic acid, acetic acid, difluoro, difluoressigsaeure, 1,1-difluoroacetic acid, unii-zqk1c95k3n, zqk1c95k3n, acetic acid, 2,2-difluoro, difluoroaceticacid, difluoracetic acid, difluoro-acetic acid
PubChem CID 9788
ChEBI CHEBI:23716
IUPAC Name 2,2-difluoroacetic acid
SMILES C(C(=O)O)(F)F

Specifications

Melting Point -1°C
Color Colorless to Brown
Density 1.5200g/mL
Boiling Point 132°C to 134°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula F2CHCO2H
Refractive Index 1.3420 to 1.345
Quantity 2.5 mL
Beilstein 02, 193
Specific Gravity 1.52
Formula Weight 96.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Difluoroacetic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.