missing translation for 'onlineSavingsMsg'
Learn More

Difluoroacetic acid, 98%

Product Code. 15720819 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 mL
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
2.5mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15720819 10 mL Glass bottle 10mL
10532761 2.5 mL Glass bottle 2.5mL
15730819 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15720819 Supplier Thermo Scientific Acros Supplier No. 162120100
  • ONLINE EXCLUSIVE PRICE
    111.00 €
    107.00 € / 10mL
    Save 4.00 €
    3% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 381-73-7 | C2H2F2O2 | 96.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 381-73-7
Molecular Formula C2H2F2O2
Molecular Weight (g/mol) 96.03
MDL Number MFCD00004220
InChI Key PBWZKZYHONABLN-UHFFFAOYSA-N
Synonym difluoroacetic acid, acetic acid, difluoro, difluoressigsaeure, 1,1-difluoroacetic acid, unii-zqk1c95k3n, zqk1c95k3n, acetic acid, 2,2-difluoro, difluoroaceticacid, difluoracetic acid, difluoro-acetic acid
PubChem CID 9788
ChEBI CHEBI:23716
IUPAC Name 2,2-difluoroacetic acid
SMILES C(C(=O)O)(F)F

Specifications

Melting Point -1°C
Color Colorless to Brown
Density 1.5200g/mL
Boiling Point 132°C to 134°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula F2CHCO2H
Refractive Index 1.3420 to 1.345
Quantity 10 mL
Beilstein 02, 193
Specific Gravity 1.52
Formula Weight 96.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Difluoroacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • Difluoroacetic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: difluoroacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.