missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Digest-All 4 (Proteinase K)

Product Code. 17156058
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17156058 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17156058 Supplier Invitrogen™ Supplier No. 003011

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product may be used to pretreat formalin-fixed, paraffin-embedded tissues (see Invitrogen Anatomical Pathology catalog page 140), and will improve the signal of certain antigens.

This product may be used to pretreat formalin-fixed, paraffin-embedded tissues (see Invitrogen Anatomical Pathology catalog page 140), and will improve the signal of certain antigens. It contains 50 mL of ready-to-use Proteinase K. Digest-All™ 4 is stable for 12 months.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.