missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ohaus™ Digital Dry Block Heater with Heated Lid
Shop All Ohaus Products
Click to view available options
No. of Blocks:
2
Unit Size:
Each
Description
- Exceptional uniformity, stability, and regulation of temperature
- These multi-purpose units are ideal for isothermal incubation, enzyme reactions, immunoassays, nucleic acid denaturation and a wide variety of other laboratory procedures
- Integral support rod holder with locking knob accepts optional External Temperature Probe Kit
- External Temperature Probe Kit monitors actual block or sample temperature
- Block heater accepts one microplate block or two separate interchangeable modular blocks (available separately) accommodating various tube sizes from 0.2 mL microtubes to test tubes or vials up to 85 mm in height
- Each block has a thermometer well for measuring block temperature
- Efficient heating due to close tube-and-block contact with a low density design for exceptional temperature uniformity
- Heated lid helps to regulate the temperature and reduce the amount of condensation on sample lids
Specifications
Specifications
| Certifications/Compliance | TUV |
| Controller Type | Digital |
| Description | Dry Block Heater, Heat Lid, HB2DGHL, EU |
| Temperature Range (Metric) | 5°C to 100°C |
| Type | Dry Bath Block |
| Heat Up Time | 50 min |
| Includes | Heated Lid |
| Model | HB2DGHL |
| No. of Blocks | 2 |
| Temperature Stability | ±0.1°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction