missing translation for 'onlineSavingsMsg'
Learn More

Aalborg Digital Mass Flowmeter, Aluminum, For Ar, 0 to 30L/min

Brand:  Aalborg XFMS-010518

223.32 EUR valid until 2026-01-16
BEST PRICE on Promo! Use promo code "24103" to get your promotional price.


Digital Mass Flowmeter, Aluminum, For Ar, 0 to 30L/min

Product Code. 16563791

  • ONLINE EXCLUSIVE PRICE
    1690.50 €
    1029.18 € / Each
    Save 661.32 €
    39% Off
Only null left
In Stock
Edge
Add to basket
This item is not returnable. View return policy

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.