Learn More
ATAGO™ Digital pH Meter DPH-2
Product Code. 11476298
Click to view available options
Range (pH):
0 to 14
Unit Size:
Each
Product Code. 11476298
Brand: ATAGO™ 4320
Only null left
Add to basket
Add to basket
Digital pH Meter DPH-2 is extremely easy to use. It has a compact, portable design and is water resistant (IP67), so it can be cleaned under running water and submersed without any negative effects. With ATC, the DPH-2 is perfect for many different applications.
  • Easy to use
  • Compact and portable
  • IP67 water resistant
  • Automatic Temperature Compensation mechanism that function from 0°C to 50°C.
  • Applications include food and beverage production control, brewing and fermentation, soil and hydroponics, cutting oils, alkali washing liquids and other soluble solutions
Specifications
Accuracy (pH) ±0.1
For Use With (Application) Food and Beverage Production Control, Brewing and Fermentation, Soil and Hydroponics, Cutting Oils, Alkali Washing Liquids, and Other Soluble Solutions
Range (pH) 0 to 14
IP Rating IP67
Model DPH-2
Power Supply LR44 Watch Battery (x4)
Resolution 0.1 pH
Temperature Compensation 0.0°C to 50.0°C