Learn More
TFA Dostmann Digital Thermo-Hygrometer
Product Code. 17901662
Change view
Click to view available options
Unit Size:
Each
Product Code. Unit Size
17901662 Each
1 options
Product Code. 17901662 Supplier TFA Dostmann Supplier No. 30.5031
  • ONLINE EXCLUSIVE PRICE
    15.02 €
    10.97 € / Pack of 1
    Save 4.05 €
    26% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
The digital thermo-hygrometer is an ideal measuring instrument for monitoring the indoor climate. It displays the room temperature, as well as the maximum and minimum values. In addition, the time or the date is shown, as well as a wake-up alarm with a snooze function can be set.
Humans spend at least two thirds of their lifetime indoors. Whether we feel good and comfortable there depends to a large extent on the quality of the air in the room. The digital thermo-hygrometer is an ideal measuring instrument for monitoring the indoor climate. You can easily keep an eye on room temperature and humidity and regulate them through targeted heating and ventilation. In this way you can create a comfortable and healthy living environment and even save on heating costs. Maximum and minimum values over a self-selected period can be called up. In addition, the thermo-hygrometer displays the time or the date, as well as a wake-up alarm with a snooze function for a gentle wake-up. The switchable hourly signal can always remind you on the hour.
  • Monitoring of indoor temperature and humidity
  • For a healthy indoor climate
  • Max.-min. values
  • Time and calendar
  • Alarm and hourly chime
Specifications
Type Digital Thermohygrometer