Learn More
TFA Dostmann Digital Thermometer
Product Code. 17757409
Change view
Click to view available options
Calibrated:
Yes
Unit Size:
Each
Product Code. Calibrated Unit Size
17757409 Yes Each
1 options
Product Code. 17757409 Supplier TFA Dostmann Supplier No. 30.1065.02
  • ONLINE EXCLUSIVE PRICE
    9.12 €
    6.90 € / Pack of 1
    Save 2.22 €
    24% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
The practical and small digital thermometer is ideal for temperature monitoring indoors. Two small magnets on the back adhere to metal surfaces. Alternatively, you can use the fold-out stand.
The practical and small thermometer is ideal for temperature monitoring indoors: in the living room, bedroom, basement, in the workshop, garage and even in the car. Two small magnets on the back adhere to metal surfaces. Alternatively, you can use the fold-out stand. Controlling the room temperature is an important factor for a comfortable and healthy living environment and it helps save energy. Even one degree less temperature can bring up to 6% reduction in heating costs.
  • Multipurpose thermometer
  • For indoor temperature measurement
  • With magnet for fixing and fold-out stand
  • Max.-min. function
  • Small and compact
  • Versatile, small and compact
Specifications
Material Plastic
Calibrated Yes
Display LCD