Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Melting Point | 240.0°C |
| Ignition Residue | 0.3% max |
| Packaging | Glass bottle |
| Quantity | 5 g |
| Merck Index | 15,318 |
| Specific Rotation Condition | −54.00 (20.00°C c=3,in methanol) |
| Specific Rotation | − 54.00 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: practically insoluble in chloroform and ether,1gr dissolves in 57mL abs. alcohol,in 220mL 95%,alcohol |
| Formula Weight | 1229.33 |
| Chemical Name or Material | Digitonin, Approx. 50% (TLC) |
Sécurité et traitement
- Digitonin
- Danger
- Acute toxicity Category 3
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.