missing translation for 'onlineSavingsMsg'
Learn More

Diglycolic acid, 98%

Product Code. 11490198
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490198 100 g 100g
11480198 25 g 25g
11400208 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490198 Supplier Thermo Scientific Alfa Aesar Supplier No. B23758.22
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-99-6 | C4H6O5 | 134.09 g/mol

Diglycolic acid was used in the preparation of needle-like aragonite particles from calcium nitrate aqueous solutions. It is also used in the synthesis of N-interlinked imipramines which show apoptotic activity against malignant cells as well as signs of combating Burkitt’s lymphoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diglycolic acid was used in the preparation of needle-like aragonite particles from calcium nitrate aqueous solutions. It is also used in the synthesis of N-interlinked imipramines which show apoptotic activity against malignant cells as well as signs of combating Burkitt’s lymphoma.

Solubility
Soluble in water, methanol.

Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-99-6
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.09
MDL Number MFCD00004309
InChI Key QEVGZEDELICMKH-UHFFFAOYSA-N
Synonym diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid
PubChem CID 8088
IUPAC Name 2-(carboxymethoxy)acetic acid
SMILES OC(=O)COCC(O)=O

Specifications

Melting Point 140°C to 145°C
Quantity 100 g
Beilstein 1072117
Solubility Information Soluble in water,methanol.
Formula Weight 134.09
Percent Purity 98%
Chemical Name or Material Diglycolic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.