missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Digoxin

Product Code. 18702624
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18702624 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18702624 Supplier R&D Systems™ Supplier No. 4583/50
  • ONLINE EXCLUSIVE PRICE
    93.00 €
    88.20 € / 50mg
    Save 4.80 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Na+/K+ ATPase inhibitor

Inhibits membrane-bound α-subunits of the Na+/K+ ATPase pump in myocytes. Increases RGS2 protein levels in primary vascular smooth muscle cells through an Na+/K+ ATPase pump-dependent mechanism. Also exhibits anti-MERS-CoV activity in Vero cells in vitro (IC50 = 0.17 μM).
TRUSTED_SUSTAINABILITY

Specifications

CAS 20830-75-5
Quantity 50 mg
Product Line Tocris
Molecular Formula C41H64O14
Formula Weight 780.94
Purity >98%
Molecular Weight (g/mol) 780.949
Chemical Name or Material Digoxin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.