missing translation for 'onlineSavingsMsg'
Learn More

Diheptyltin Dichloride, TRC

Product Code. 30138077
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138077 1 g 1g
30011544 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138077 Supplier Toronto Research Chemicals Supplier No. TRCD4506881G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74340-12-8
Molecular Formula C14 H30 Cl2 Sn
Molecular Weight (g/mol) 388
Synonym Dichlorodiheptylstannane, Diheptyltin dichloride, Di-n-heptyltin dichloride
IUPAC Name dichloro(diheptyl)stannane
SMILES CCCCCCC[Sn](Cl)(Cl)CCCCCCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/2C7H15.2ClH.Sn/c2*1-3-5-7-6-4-2;;;/h2*1,3-7H2,2H3;2*1H;/q;;;;+2/p-2
Formula Weight 388.0747
Chemical Name or Material Di-n-heptyltin-dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.