missing translation for 'onlineSavingsMsg'
Learn More

Dihexylamine, 99+%

Product Code. 10548892 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10548892 100 g Glass bottle 100g
10538892 5 g Glass bottle 5g
10249573 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10548892 Supplier Thermo Scientific Acros Supplier No. 407591000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 143-16-8 | C12H27N | 185.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 143-16-8
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.35
MDL Number MFCD00009521
InChI Key PXSXRABJBXYMFT-UHFFFAOYSA-N
Synonym dihexylamine, di-n-hexylamine, 1-hexanamine, n-hexyl, n,n-dihexylamine, n-hexylhexanamine, unii-k37ada14zv, ccris 4622, k37ada14zv, di-n-heylamine, bis 1-hexyl amine
PubChem CID 8920
IUPAC Name N-hexylhexan-1-amine
SMILES CCCCCCNCCCCCC

Specifikationer

Melting Point 3°C
Color Colorless
Density 0.8000g/mL
Boiling Point 236°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula (CH3(CH2)5)2NH
Refractive Index 1.4320 to 1.434
Quantity 100 g
UN Number 2927
Beilstein 04,I,384
Specific Gravity 0.8
Solubility Information Solubility in water: 0.3g/L(25°C)
Formula Weight 185.35
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Dihexylamine
Visa mer Visa mindre
compliance-icons compliance-icons
Produktbeteckning
  • Dihexylamine
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Faroangivelser
  • H310-Fatal in contact with skin.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.