missing translation for 'onlineSavingsMsg'
Learn More

Dihydro Ferulic Acid 4-O-beta-D-Glucuronide, TRC

Product Code. 30132739
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132739 25 mg 25mg
30005483 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132739 Supplier Toronto Research Chemicals Supplier No. TRCD44831525MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86321-28-0
Molecular Formula C16H20O10
Molecular Weight (g/mol) 372.32
Synonym 4-(2-Carboxyethyl)-2-methoxyphenyl β-D-Glucopyranosiduronic Acid, Dihydroferulic Acid 4-O-Glucuronide
SMILES COc1cc(CCC(=O)O)ccc1O[C@H]2O[C@H]([C@H](O)[C@@H](O)[C@@H]2O)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H20O10/c1-24-9-6-7(3-5-10(17)18)2-4-8(9)25-16-13(21)11(19)12(20)14(26-16)15(22)23/h2,4,6,11-14,16,19-21H,3,5H2,1H3,(H,17,18)(H,22,23)/t11-,12-,13+,14-,16+/m0/s1
Percent Purity >95%
Chemical Name or Material Dihydro Ferulic Acid 4-O-Beta-D-Glucuronide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.