missing translation for 'onlineSavingsMsg'
Learn More

Dihydro Ferulic Acid, TRC

Product Code. 30373138
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30373138 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373138 Supplier Toronto Research Chemicals Supplier No. TRCD448900500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroferulic Acid
CAS 1135-23-5
Molecular Formula C10 H12 O4
Quantity 500 mg
Synonym Benzenepropanoic acid, 4-hydroxy-3-methoxy-, Hydrocinnamic acid, 4-hydroxy-3-methoxy- (6CI,7CI,8CI), 4-Hydroxy-3-methoxybenzenepropanoic acid, (4-Hydroxy-3-methoxyphenyl)propionic acid, 3-(3-Methoxy-4-hydroxyphenyl)propanoic acid, 3-(3-Methoxy-4-hydroxyphenyl)propionic acid, 3-(3'-Methoxy-4'-hydroxyphenyl)propionic acid, 3-(4-Hydroxy-3-methoxyphenyl)propanoic acid, 3-(4-Hydroxy-3-methoxyphenyl)propionic acid, 3-Methoxy-4-hydroxyphenylpropionic acid, 3-Methoxyphloretic acid, Dihydroconiferylic acid, Dihydroferulic acid, Ferulic acid, α,β-dihydro-, Hydroferulic acid, Shorbic acid, β-(4-Hydroxy-3-methoxyphenyl)propionic acid, β-3-Methoxy-4-hydroxyphenylpropionic acid
InChI Formula InChI=1S/C10H12O4/c1-14-9-6-7(2-4-8(9)11)3-5-10(12)13/h2,4,6,11H,3,5H2,1H3,(H,12,13)
SMILES COc1cc(CCC(=O)O)ccc1O
IUPAC Name 3-(4-hydroxy-3-methoxyphenyl)propanoic acid
Molecular Weight (g/mol) 196.2
Formula Weight 196.0736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.