missing translation for 'onlineSavingsMsg'
Learn More

Dihydroergotamine Mesylate, TRC

Product Code. 30384033
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384033 5 mg 5mg
30378796 100 mg -
30377314 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384033 Supplier Toronto Research Chemicals Supplier No. TRCD4490805MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6190-39-2
Molecular Formula C33 H37 N5 O5 . C H4 O3 S
Molecular Weight (g/mol) 679.78
Synonym Dihydroergotamine mesilate, (6aR,9R,10aR)-N-[(2R,5S,10aS,10bS)-5-Benzyl-10b-hydroxy-2-methyl-3,6-dioxooctahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl]-7-methyl-4,6,6a,7,8,9,10,10a- octahydroindolo[4,3-fg]quinoline-9-carboxamide methanesulfonate, Ergotaman-3',6',18-trione, 9,10-dihydro-12'-hydroxy-2'-methyl-5'-(phenylmethyl)-, (5'α,10α)-, methanesulfonate (1:1), Ergotaman-3',6',18-trione, 9,10-dihydro-12'-hydroxy-2'-methyl-5'-(phenylmethyl)-, (5'α,10α)-, monomethanesulfonate (salt) (9CI), Ergotamine, 9,10-dihydro-, monomethanesulfonate (salt) (6CI,7CI,8CI), (+)-Dihydroergotamine mesylate, 9,10-Dihydroergotamine mesylate, Agit, Angionorm, DETMS, DHE 45, Dergotamine, Diergo, Dihydergot, Dihydroergotamine mesylate, Dihydroergotamine methanesulfonate, Dihydroergotamine methanesulphonate, Dihydroergotamine monomethanesulfonate, Dihytamine, Dirgotarl, Endophleban, Ergomimet, Ergont, Ergotonin, Ikaran, Migranal, Morena, Orstanorm, Seglor, Tonopres, Verladyn
IUPAC Name (6aR,9R,10aR)-N-[(2R,5S,10aS,10bS)-5-Benzyl-10b-hydroxy-2-methyl-3,6-dioxooctahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl]-7-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinoline-9-carboxamide methanesulfonate
SMILES CN1C[C@@H](C[C@H]2[C@H]1Cc3c[nH]c4cccc2c34)C(=O)N[C@]5(C)O[C@@]6(O)[C@@H]7CCCN7C(=O)[C@H](Cc8ccccc8)N6C5=O.CS(=O)(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C33H37N5O5.CH4O3S/c1-32(35-29(39)21-15-23-22-10-6-11-24-28(22)20(17-34-24)16-25(23)36(2)18-21)31(41)38-26(14-19-8-4-3-5-9-19)30(40)37-13-7-12-27(37)33(38,42)43-32;1-5(2,3)4/h3-6,8-11,17,21,23,25-27,34,42H,7,12-16,18H2,1-2H3,(H,35,39);1H3,(H,2,3,4)/t21-,23-,25-,26+,27+,32-,33+;/m1./s1
Formula Weight 679.2676
Chemical Name or Material Dihydroergotamine mesilate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.