missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen Hexachloroplatinate Hexahydrate, TRC

Product Code. 30377361
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30377361 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30377361

Brand: Toronto Research Chemicals TRCD4506305G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dihydrogen Hexachloroplatinate(IV) hexahydrate
CAS 18497-13-7
Molecular Formula Cl6 Pt H2 . 6 H2 O
Quantity 5 g
Synonym Chloroplatinic Acid Hexahydrate, Dihydrogen Hexachloroplatanate(2-) Hexahydrate, Dihydrogen hexachloroplatinate hexahydrate, Dihydrogen hexachloroplatinate(2-) hexahydrate, Hexachloroplatinic acid hexahydrate, NSC 4958
InChI Formula InChI=1S/6ClH.6H2O.Pt/h6*1H;6*1H2;/q;;;;;;;;;;;;+4/p-4
SMILES O.O.O.O.O.O.Cl[Pt-2](Cl)(Cl)(Cl)(Cl)Cl
IUPAC Name hexachloroplatinum(2-);hydron;hexahydrate
Molecular Weight (g/mol) 517.9
Formula Weight 514.8569
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.