missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexafluorozirconate, 20% w/w aq. soln., Thermo Scientific Chemicals

Product Code. 11318268
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318268 5 kg 5kg
11308268 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318268 Supplier Thermo Scientific Alfa Aesar Supplier No. 039461.A7
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12021-95-3
Molecular Formula F6H2Zr
Molecular Weight (g/mol) 207.23
MDL Number MFCD00082965
InChI Key PZFQYBOZXGWNAM-UHFFFAOYSA-H

Specifications

Color Green
Density 1.52 g/mL
Quantity 5 kg
UN Number UN3264
Formula Weight 207.22
Concentration or Composition (by Analyte or Components) 20% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Dihydrogen hexafluorozirconate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.