missing translation for 'onlineSavingsMsg'
Learn More

Dihydrostreptomycin sulfate, MedChemExpress

Product Code. 16492319
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492319 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16492319 Supplier Medchem Express Supplier No. HYB1241
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dihydrostreptomycin sulfate is an aminoglycoside antibiotic, used to treat bacterial diseases in cattle, pigs and sheep.

Dihydrostreptomycin sulfate is an aminoglycoside antibiotic, used to treat bacterial diseases in cattle, pigs and sheep.

Specifications

Chemical Name or Material Dihydrostreptomycin sulfate
CAS 5490-27-7
Color White
Purity Grade Notes Research
Molecular Formula C21H41N7O12.3/2H2SO4
Quantity 1 g
Synonym Dihydrostreptomycin sesquisulfate
Solubility Information H2O : ≥ 200 mg/mL (273.71 mM)
SMILES O[C@@H]1[C@@H](O)[C@@](O[C@@H]2O[C@@H](C)[C@](O)(CO)[C@H]2O[C@@]3([H])[C@@H](NC)[C@H](O)[C@@H](O)[C@H](CO)O3)([H])[C@@H](NC(N)=N)[C@H](O)[C@H]1NC(N)=N.[1.5H2SO4]
Molecular Weight (g/mol) 730.71
Formula Weight 730.71
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.