missing translation for 'onlineSavingsMsg'
Learn More

Dihydrouracil, MedChemExpress

Product Code. 16439366
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16439366 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16439366 Supplier Medchem Express Supplier No. HYW012926
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dihydrouracil (5,6-Dihydrouracil), a metabolite of Uracil, can be used as a marker for identification of dihydropyrimidine dehydrogenase (DPD)-deficient.

Dihydrouracil (5,6-Dihydrouracil), a metabolite of Uracil, can be used as a marker for identification of dihydropyrimidine dehydrogenase (DPD)-deficient.

Specifications

Chemical Name or Material Dihydrouracil
CAS 504-07-4
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C4H6N2O2
Quantity 100 mg
Synonym 5,6-Dihydrouracil
Solubility Information DMSO : 14.29 mg/mL (125.24 mM; Need ultrasonic)
SMILES O=C1CCNC(N1)=O
Molecular Weight (g/mol) 114.1
Formula Weight 114.1
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.