missing translation for 'onlineSavingsMsg'
Learn More

Diiodomethane, 99%, stab.

Product Code. 11468790
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
500 g
1000 g
Unit Size:
1000g
100g
250g
25g
500g
50g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468790 100 g 100g
11458790 25 g 25g
11432817 50 g 50g
11442817 250 g 250g
11478790 500 g 500g
11422817 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11468790 Supplier Thermo Scientific Alfa Aesar Supplier No. A15457.22
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-11-6 | CH2I2 | 267.84 g/mol

Diiodomethane is used to determine the density of minerals and other solid samples due to its high density. It is used as an optical contact liquid to determine the refractive index of certain gemstones. In simmons-smith reaction, it acts as a reagent to generate methylene (carbine) free radical. It reacts with olefins to prepare cyclopropanes with high stereo specificity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diiodomethane is used to determine the density of minerals and other solid samples due to its high density. It is used as an optical contact liquid to determine the refractive index of certain gemstones. In simmons-smith reaction, it acts as a reagent to generate methylene (carbine) free radical. It reacts with olefins to prepare cyclopropanes with high stereo specificity.

Solubility
Miscible with alcohol and ether. Slightly miscible with water.

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with alkali metal salts, strong oxidizing agents, strong bases, metals, lithium, potassium, sodium, sodium oxides and alloys.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-11-6
Molecular Formula CH2I2
Molecular Weight (g/mol) 267.84
MDL Number MFCD00001079
InChI Key NZZFYRREKKOMAT-UHFFFAOYSA-N
Synonym methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
PubChem CID 6346
IUPAC Name diiodomethane
SMILES ICI

Specifications

Melting Point 5°C to 6°C
Density 3.322
Boiling Point 180°C to 181°C (decomposition)
Flash Point None
Assay Percent Range 99%, stab.
Refractive Index 1.74
Quantity 100 g
UN Number UN2810
Beilstein 1696892
Sensitivity Light sensitive
Merck Index 14,6066
Solubility Information Miscible with alcohol and ether. Slightly miscible with water.
Formula Weight 267.84
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diiodomethane, stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diiodomethane, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Methylene iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.