missing translation for 'onlineSavingsMsg'
Learn More

Diiron nonacarbonyl, 98%

Product Code. 10398150 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10398150 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10398150 Supplier Thermo Scientific Acros Supplier No. 209280500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 15321-51-4 | C9H3Fe2O9 | 366.804 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15321-51-4
Molecular Formula C9H3Fe2O9
MDL Number MFCD00151465
InChI Key CGJSNKHFIBINMR-UHFFFAOYSA-N
Synonym 1,3-diferrabicyclo 1.1.1 pentane-2,4,5-trione; hexakis carbon monoxide, diironnonacarbonyl, purum fe, nonacarbonyldiiron, iron nonacarbonyl, diironnonacarbonyl, enneacarbonyldiiron
PubChem CID 71311424

Specifications

Color Black to Brown-Red
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula Fe2(CO)9
Quantity 50 g
Fieser 01,259; 02,139; 03,101; 04,157; 05,221; 06,195; 07,110; 08,498; 13,320; 04,534; 05,706; 07,144; 08,41; 08,216; 10,174; 08,214
SMILES [Fe].[Fe+3].[CH-]=O.[CH-]=O.[CH-]=O.[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
IUPAC Name iron(3+) hexakis(methanidylidyneoxidanium) tris(oxomethanide) iron
Molecular Weight (g/mol) 366.80
Formula Weight 363.78
Percent Purity ≥97.5%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Diiron nonacarbonyl, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.