missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylaluminum chloride, 98+%, 25% w/w in hexane, Thermo Scientific™

Product Code. 11396389
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11396389 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11396389 Supplier Thermo Scientific Alfa Aesar Supplier No. 089039.NF
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Catalyst component in Ziegler-Natta type catalyst systems; in alkylation reactions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1779-25-5
Molecular Formula C8H18AlCl
Molecular Weight (g/mol) 176.664
MDL Number MFCD00008927
InChI Key HQMRIBYCTLBDAK-UHFFFAOYSA-M
Synonym diisobutylaluminum chloride, chlorodiisobutylaluminium, chlorodiisobutylaluminum, diisobutylchloroaluminum, aluminum, chlorobis 2-methylpropyl, aluminum, chlorodiisobutyl, bis isobutyl aluminum chloride, diisobutylaluminum monochloride, chlorobis 2-methylpropyl aluminum, alluminio diisobutil-monocloruro italian
PubChem CID 15688
IUPAC Name chloro-bis(2-methylpropyl)alumane
SMILES CC(C)C[Al](CC(C)C)Cl

Specifications

Color Colorless
Density 0.72 g/mL
Linear Formula [(CH3)2CHCH2]2AlCl
Quantity 100 g
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information Soluble in aromatic,saturated aliphatic and cycloaliphatic hydrocarbon. Reacts with water.
Formula Weight 176.67
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Diisobutylaluminum chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.