missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylaluminum hydride, 1M solution in hexane, Thermo Scientific™

Product Code. 15421827
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15421827 1 L 1 L
15416495 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 15421827 Supplier Thermo Scientific Alfa Aesar Supplier No. 042589.K2
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Reducing agent

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-15-7
MDL Number MFCD00008928

Specifications

Color Colorless
Density 0.701 g/mL
Flash Point −23°C (−10°F)
Linear Formula [(CH3)2CHCH2]2AlH
Quantity 1 L
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information Miscible with hydrocarbon solvents.
Formula Weight 142.22
Physical Form Liquid
Chemical Name or Material Diisobutylaluminum hydride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.