missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylaluminum hydride, 1M solution in toluene, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15464875
Change view
Click to view available options
Quantity:
50 mL
100 mL
200 mL
500 mL
Unit Size:
100mL
200mL
500mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464875 500 mL 500mL
11369208 50 mL 50mL
15454875 100 mL 100mL
11359208 200 mL 200mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 15464875 Supplier Thermo Scientific Alfa Aesar Supplier No. 042591.AP
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-15-7
MDL Number MFCD00008928

Specifications

Color Colorless
Density 0.805 g/mL
Boiling Point ∼110°C
Flash Point 4°C (40°F)
Packaging ChemSeal™ bottles
Linear Formula [(CH3)2CHCH2]2AlH
Quantity 500 mL
UN Number UN3399
Beilstein 4123663
Sensitivity Air and moisture sensitive
Solubility Information Miscible with organic solvents.
Formula Weight 142.22
Physical Form Liquid
Chemical Name or Material Diisobutylaluminum hydride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.