missing translation for 'onlineSavingsMsg'
Learn More

Diisobutylaluminum hydride, 25% w/w in hexane, Thermo Scientific™

Product Code. 15491817
Change view
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491817 50 mL 50mL
15401827 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 15491817 Supplier Thermo Scientific Alfa Aesar Supplier No. 042588.AD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Reducing agent

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-15-7
MDL Number MFCD00008928
Synonym DIBAL-H

Specifications

Color Colorless
Density 0.701g/mL
Linear Formula [(CH3)2CHCH2]2AlH
Quantity 50 mL
UN Number UN3399
Sensitivity Air & Moisture Sensitive
Formula Weight 142.22
Physical Form Liquid
Chemical Name or Material Diisobutylaluminum hydride
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diisobutylaluminum hydride, 25% w/w in hexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Substance which, in contact with water, emits flammable gases Category 1
  • Skin corrosion/irritation Category 1 B
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H336-May cause drowsiness or dizziness.
  • H373-May cause damage to organs.
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.