missing translation for 'onlineSavingsMsg'
Learn More

Diisopropanolamine, 99%

Product Code. 10259640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10259640 500 g Glass bottle 500g
10687274 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10259640 Supplier Thermo Scientific Acros Supplier No. 407805000
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-97-4 | C6H15NO2 | 133.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 110-97-4
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00004531
InChI Key LVTYICIALWPMFW-UHFFFAOYNA-N
Synonym diisopropanolamine, 1,1'-iminodipropan-2-ol, bis 2-hydroxypropyl amine, 2-propanol, 1,1'-iminobis, bis 2-propanol amine, 1,1'-iminodi-2-propanol, dipa alcohol, di-2-propanolamine, 1,1'-azanediylbis propan-2-ol, dipropyl-2,2'-dihydroxy-amine
PubChem CID 8086
IUPAC Name 1-[(2-hydroxypropyl)amino]propan-2-ol
SMILES CC(O)CNCC(C)O

Tekniske data

Melting Point 33°C
Color Colorless to Yellow
Density 1.0038g/mL
Boiling Point 249°C to 250°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 98.5 % min. (Total base)
Packaging Glass bottle
Linear Formula (CH3CHOHCH2)2NH
Refractive Index 1.4615 to 1.4635
Quantity 500 g
Merck Index 15, 3214
Specific Gravity 1.0038
Solubility Information Solubility in water: 870g/L (20°C)
Viscosity 84.19 mPa.s (60°C)
Formula Weight 133.19
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material Diisopropanolamine, 99%
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • Diisopropanolamine
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
Faresætning
  • H319-Causes serious eye irritation.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.