missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl azodicarboxylate, 94%

Product Code. 10091781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10091781 100 g 100g
10570812 5 g 5g
10329040 25 g 25g
15194027 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10091781 Supplier Thermo Scientific Acros Supplier No. 327561000
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2446-83-5 | C8H14N2O4 | 202.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X Bond Formation (Halogen), Radical initiators
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2446-83-5
Molecular Formula C8H14N2O4
Molecular Weight (g/mol) 202.21
MDL Number MFCD00008875
InChI Key VVWRJUBEIPHGQF-KTKRTIGZSA-N
Synonym z-n-propan-2-yloxy carbonyl imino propan-2-yloxy formamide
PubChem CID 56776520
IUPAC Name propan-2-yl (NZ)-N-propan-2-yloxycarbonyliminocarbamate
SMILES CC(C)OC(=O)N=NC(=O)OC(C)C

Specifications

Melting Point 3.0°C to 5.0°C
Color Orange-Yellow to Red
Density 1.0200g/mL
Boiling Point 75.0°C (0.2 mmHg)
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHO2CN=NCO2CH(CH3)2
Refractive Index 1.4180 to 1.4220
Quantity 100 g
Fieser 17,39
Merck Index 15,3217
Specific Gravity 1.02
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 202.21
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Diisopropyl azodicarboxylate, 94%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diisopropyl azodicarboxylate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.