missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Azodicarboxylate, TRC

Product Code. 30375733
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30375733 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375733 Supplier Toronto Research Chemicals Supplier No. TRCD45524010G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diisopropyl Azodicarboxylate
CAS 2446-83-5
Purity Grade Notes HPLC
Molecular Formula C8H14N2O4
Quantity 10 g
Synonym 1,2-Diazenedicarboxylic Acid 1,2-Bis(1-methylethyl) Ester, Azodiformic Acid Diisopropyl Ester, DIAD, Diisopropyl Azodiformate, Diisopropyl diazodicarboxylate
InChI Formula InChI=1S/C8H14N2O4/c1-5(2)13-7(11)9-10-8(12)14-6(3)4/h5-6H,1-4H3/b10-9+
SMILES CC(C)OC(=O)N=NC(=O)OC(C)C
Molecular Weight (g/mol) 202.21
Formula Weight 202.0954
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.