missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl ether, Solstice™

Product Code. 16696163
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
16696163 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16696163 Supplier Honeywell Riedel-de Haën Supplier No. 3982761L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, contains either BHT or hydroquinone as stabilizer, ≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Specifications

CAS 108-20-3
Boiling Point 67°C to 70°C (1.013 hPa)
Flash Point -29 °C
Packaging Glass Bottle
Molecular Formula C6H14O
Quantity 1 L
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
SMILES CC(C)OC(C)C
IUPAC Name 2-(propan-2-yloxy)propane
Molecular Weight (g/mol) 102.18
PubChem CID 7914
Percent Purity min. 99.0 %
Grade ACS General Use Solvents
Physical Form Liquid
Show More Show Less
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.