missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl ether, puriss p.a., ≥98.5% (GC), Solstice

Product Code. 15655260
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15655260 1 L Glass bottle 1 L
15665260 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15655260 Supplier Fluka Supplier No. 382701L
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Puriss. p.a.,≥98.5% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-20-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00008880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Specifications

Melting Point -86°C
Density 0.724g/mL at 20°C, 0.725g/mL at 25°C
Boiling Point 67°C to 70°C
Flash Point −29°C (−20.2°F)
Packaging Glass bottle
Linear Formula (CH3)2CHOCH(CH3)2
Refractive Index 1.367
Quantity 1 L
UN Number UN1159
Beilstein 1731256
Vapor Density 3.5 (vs air)
Formula Weight 102.17g/mol
Percent Purity ≥98.5% (GC)
Chemical Name or Material Diisopropyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diisopropyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.