missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Ether, ReagentPlus™, 99%, Honeywell™

Product Code. 15631670 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
Packaging:
Aluminum Bottle
Glass Bottle
Unit Size:
1 L
2.5 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15631670 1 L Glass Bottle 1 L
15641670 2.5 L Glass Bottle 2.5 L
15618330 4 L Aluminum Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15631670 Supplier Honeywell Research Chemicals Supplier No. 1853021L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, contains either BHT or hydroquinone as stabilizer, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-20-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00008880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Specifications

Melting Point -86°C
Color Colorless
Density 0.725g/mL at 25°C
Boiling Point 70°C
Explosion Limit 1 to 21% (100°F)
Flash Point −29°C (−20.2°F)
Ignition Point 827°F
Packaging Glass Bottle
Linear Formula (CH3)2CHOCH(CH3)2
Refractive Index n20/D 1.367(lit.)
Quantity 1 L
UN Number UN1159
Beilstein 1731256
Vapor Density 3.5 (vs air)
Formula Weight 102.17g/mol
Vapor Pressure 120mmHg (20°C)
Percent Purity 99%
Grade ReagentPlus
Physical Form Liquid
Chemical Name or Material Diisopropyl Ether, Contains either BHT or hydroquinone as stabilizer
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diisopropyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.