missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (-)-Diisopropyl-L-malate, 97%

Product Code. p-7026134
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11455067

Brand: Alfa Aesar™ B21960.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 83541-68-8
Molecular Formula C10H16O5
Molecular Weight (g/mol) 216.23
MDL Number MFCD00274300
InChI Key DQOODBSVDOAUMP-JTQLQIEISA-L
Synonym diisopropyl s---malate, --diisopropyl-l-malate, 1,4-diisopropyl 2s-2-hydroxybutanedioate, diisopropyl d-malate, s-diisopropyl malate, diisopropyl-s---malate, l-malic acid diisopropyl ester, s-2-hydroxybutanedioic acid diisopropyl ester
PubChem CID 11333435
SMILES CC(C)O[C@@](CC([O-])=O)(C(C)C)C([O-])=O

Specifications

Density 1.055
Boiling Point 236°C to 238°C
Quantity 25 g
IUPAC Name (2S)-2-(propan-2-yl)-2-(propan-2-yloxy)butanedioate
Formula Weight 218.25
Percent Purity 97%
Chemical Name or Material (-)-Diisopropyl-L-malate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.