missing translation for 'onlineSavingsMsg'
Learn More

(+)-Diisopropyl L-tartrate, 99%, ACROS Organics™

Product Code. 10134840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10134840 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10134840 Supplier Thermo Scientific Acros Supplier No. 212030250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 2217-15-4
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.11g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C10H18O6
Linear Formula {-CH(OH)CO2CH(CH3)2}2
Refractive Index 1.4380 to 1.4400 (20°C, 589nm)
MDL Number MFCD00064450
Quantity 25 g
Beilstein 03, 517
Fieser 13,276; 16,339; 12,493; 13,310
Specific Rotation Condition +17.25° (20°C neat)
Specific Rotation +16° to +18.5° (20°C, 589nm) (neat)
Synonym +-diisopropyl l-tartrate, 2r,3r-diisopropyl 2,3-dihydroxysuccinate, di-isopropyl tartrate, diisoprropyl-l-tartrate, +-dipt, l-+-tartaric acid diisopropyl ester, diisopropyl l-tartrate, l-+-diisopropyl-tartrate, l-dipt, butanedioic acid, 2,3-dihydroxy-, bis 1-methylethyl ester, r,r-+/-, 1,4-diisopropyl 2r,3r-2,3-dihydroxybutanedioate
InChI Key XEBCWEDRGPSHQH-HTQZYQBOSA-N
SMILES CC(C)OC(=O)C(C(C(=O)OC(C)C)O)O
IUPAC Name dipropan-2-yl (2R,3R)-2,3-dihydroxybutanedioate
Molecular Weight (g/mol) 234.248
PubChem CID 6453580
Formula Weight 234.25
Percent Purity ≥98.5%
Physical Form Viscous Liquid
Chemical Name or Material (+)-Diisopropyl L-tartrate, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.