missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl phosphite, 98%

Product Code. 11480258
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480258 100 g 100g
11470258 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480258 Supplier Thermo Scientific Alfa Aesar Supplier No. B23800.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1809-20-7 | C6H14O3P- | 165.149 g/mol

Diisopropyl phosphite is used as antiwear lubricant additive together with triphenyl thiophosphate on T8 steel/Al2O3 ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diisopropyl phosphite is used as antiwear lubricant additive together with triphenyl thiophosphate on T8 steel/Al2O3 ceramics.

Solubility
Soluble in water, chloroform.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1809-20-7
Molecular Formula C6H14O3P-
Molecular Weight (g/mol) 165.149
MDL Number MFCD00117905
InChI Key IWEDBEZKWHRUJX-UHFFFAOYSA-N
Synonym diisopropylphosphite, acmc-1bsaz, phosphorous acid diisopropyl anion, phosphorous acid, bis 1-methylethyl ester
PubChem CID 21884976
IUPAC Name dipropan-2-yl phosphite
SMILES CC(C)OP([O-])OC(C)C

Specifications

Density 0.99
Boiling Point 72°C to 75°C (10 mmHg)
Flash Point 112°C (234°F)
Linear Formula [(CH3)2CHO]2P(O)H
Refractive Index 1.4075
Quantity 100 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water,chloroform.
Formula Weight 166.16
Percent Purity 98%
Chemical Name or Material Diisopropyl phosphite
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.