missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Phosphite, TRC

Product Code. 30374774
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30374774 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30374774 Supplier Toronto Research Chemicals Supplier No. TRCD45937010G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diisopropyl Phosphite
CAS 1809-20-7
Molecular Formula C6H15O3P
Quantity 10 g
Synonym Diisopropyl Phosphonate, Phosphonic Acid Diisopropyl Ester, Phosphorous Acid Diisopropyl Ester, Phosphonic Acid Bis(1-methylethyl) Ester, Diisopropoxyphosphine Oxide, Diisopropyl Hydrogen Phosphite, O,O-Diisopropyl Phosphonate
InChI Formula InChI=1S/C6H15O3P/c1-5(2)8-10(7)9-6(3)4/h5-7H,1-4H3
SMILES CC(C)OP(O)OC(C)C
IUPAC Name dipropan-2-yl hydrogen phosphite
Molecular Weight (g/mol) 166.16
Formula Weight 166.0759
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.