missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylamine 2,2-Dichloroacetate, TRC

Product Code. 30134726
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134726 10 g 10g
30133150 1 g 1g
17918719 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30134726 Supplier Toronto Research Chemicals Supplier No. TRCD07271510G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 660-27-5
Molecular Formula C6H15N . C2H2Cl2O2
Molecular Weight (g/mol) 230.13
Synonym Cubisol, DADA, DIEDI, DIPA, DIPA-DCA, Dapocel, Dedyl Diisopropylamine Dichloroethanoate, Diisopropylammonium Dichloroacetate, Diisopropylammonium Dichloroethanoate, Dipromonium, Disotat, IS 401, Kalodil, Krino B 15, Oxypangam, Tensicor, Vasculopatina, beta-Anoxin;
IUPAC Name 2,2-dichloroacetic acid;N-propan-2-ylpropan-2-amine
SMILES CC(C)NC(C)C.OC(=O)C(Cl)Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H15N.C2H2Cl2O2/c1-5(2)7-6(3)4;3-1(4)2(5)6/h5-7H,1-4H3;1H,(H,5,6)
Formula Weight 229.0636
Chemical Name or Material Diisopropylamine 2,2-Dichloroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.